KEY428412364 Key Organics / BIONET

methyl 3-(4-methoxyphenyl)-3-methylbutanoate

  • MDL number MFCD22688506
  • Cas Number : 67437-37-0
  • Molecular Weight : 222.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 0.90
Storage temp. RT
Physical Form Colourless to Yellow Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437043

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4272_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart