KEY428412369 Key Organics / BIONET

1-hydroxy-2,3-dihydro-1H-indene-5-carbonitrile

  • MDL number MFCD14708197
  • Cas Number : 125114-88-7
  • Molecular Weight : 159.188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to Grey Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437049

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4288_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart