KEY428412374 Key Organics / BIONET

2,5-dibromo-4-nitropyridin-1-ium-1-olate

  • MDL number MFCD00233996
  • Cas Number : 221241-25-4
  • Molecular Weight : 297.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437054

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4299_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart