KEY428412375 Key Organics / BIONET

N-(4-bromo-2-methoxyphenyl)-2,2,2-trifluoroacetamide

  • MDL number MFCD28101534
  • Cas Number : 870838-52-1
  • Molecular Weight : 298.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437055

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4300_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart