KEY428412378 Key Organics / BIONET

tert-butyl 6-methoxy-5-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

  • MDL number MFCD28101535
  • Cas Number : 921224-63-7
  • Molecular Weight : 308.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437058

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4307_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart