KEY428412379 Key Organics / BIONET

5-iodo-4-methoxy-2-(propan-2-yl)phenyl 4-methylbenzene-1-sulfonate

  • MDL number MFCD20486951
  • Cas Number : 865305-64-2
  • Molecular Weight : 446.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437059

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4308_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart