KEY428412381 Key Organics / BIONET

1-bromo-2,4-dimethyl-3-nitrobenzene

  • MDL number MFCD00092653
  • Cas Number : 39053-43-5
  • Molecular Weight : 230.061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437061

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4310_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart