KEY428412384 Key Organics / BIONET

3-(benzenesulfonyl)-8-nitroquinoline

  • MDL number MFCD09261163
  • Cas Number : 607743-07-7
  • Molecular Weight : 314.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437064

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4313_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart