KEY428412389 Key Organics / BIONET

5-fluoro-6-methoxy-2,3-dihydro-1H-inden-1-one

  • MDL number MFCD09261202
  • Cas Number : 83802-71-5
  • Molecular Weight : 180.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-6-methoxy-1-indanone
Inchi Code 1S/C10H9FO2/c1-13-10-5-7-6(4-8(10)11)2-3-9(7)12/h4-5H,2-3H2,1H3
Inchi Key FFZRFPIFJVKURP-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437069

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4319_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart