KEY428412392 Key Organics / BIONET

tert-butyl 6-methoxy-7-nitro-1,2,3,4-tetrahydroisoquinoline-2-carboxylate

  • MDL number MFCD28101536
  • Cas Number : 921224-62-6
  • Molecular Weight : 308.334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437072

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4326_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart