KEY428412398 Key Organics / BIONET

2-(dimethylamino)-4-fluoro-3-formylbenzonitrile

  • MDL number MFCD28101537
  • Cas Number : 1515395-53-5
  • Molecular Weight : 192.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Yellow to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437078

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4336_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart