KEY428412406 Key Organics / BIONET

1-cyclobutyl-4-(piperidine-4-carbonyl)piperazine

  • MDL number MFCD09261268
  • Cas Number : 799559-59-4
  • Molecular Weight : 251.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437086

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4348_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart