KEY428412408 Key Organics / BIONET

benzyl (3S)-3-methylpiperazine-1-carboxylate

  • MDL number MFCD04039319
  • Cas Number : 612493-87-5
  • Molecular Weight : 234.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp LMS
Iupac Name benzyl (3S)-3-methyl-1-piperazinecarboxylate
Inchi Code 1S/C13H18N2O2/c1-11-9-15(8-7-14-11)13(16)17-10-12-5-3-2-4-6-12/h2-6,11,14H,7-10H2,1H3/t11-/m0/s1
Inchi Key JRPIQMPFKMFAOX-NSHDSACASA-N

Description

Melting Point LMS
Purity 90%
Storage temp. Store at 4�Light sensitive
Physical Form Low Melting Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437088

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404,P410,P411
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4350_SDS.pdf
hcodes H302,H312,H315,H319,H332,H335
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart