KEY428412410 Key Organics / BIONET

N'-(oxolan-3-ylidene)(tert-butoxy)carbohydrazide

  • MDL number MFCD28101538
  • Cas Number : 158020-60-1
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437090

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4421_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart