KEY428412415 Key Organics / BIONET

2-chloro-5-nitro-1,3-benzoxazole

  • MDL number MFCD11111726
  • Cas Number : 54120-91-1
  • Molecular Weight : 198.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 172 - 175
Purity 97
Storage temp. Store Under N2
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437099

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/FK-0723_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart