KEY428412416 Key Organics / BIONET

(S)-N-[(1E)-ethylidene]-2-methylpropane-2-sulfinamide

  • MDL number MFCD18781421
  • Cas Number : 623168-99-0
  • Molecular Weight : 147.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437100

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GK-0705_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart