KEY428412418 Key Organics / BIONET

1,6-Anhydro-2-Deoxy-2-iodo-d-glucopyranose

  • MDL number MFCD14636511
  • Cas Number : 139437-39-1
  • Molecular Weight : 272.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 105 �107
Purity 0.97
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437102

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GK-0711_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart