KEY428412420 Key Organics / BIONET

N-[bis(methylsulfanyl)methylidene]-4-chlorobenzene-1-sulfonamide

  • MDL number MFCD28101539
  • Cas Number : 13068-12-7
  • Molecular Weight : 295.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437462

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-3408_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart