KEY428412422 Key Organics / BIONET

(6S)-6-(propylamino)-5,6,7,8-tetrahydronaphthalen-1-ol hydrobromide

  • MDL number MFCD20485837
  • Cas Number : 165950-84-5
  • Molecular Weight : 286.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437464

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-3533_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart