KEY428412429 Key Organics / BIONET

(4R)-3-(2-bromoacetyl)-4-phenyl-1,3-oxazolidin-2-one

  • MDL number MFCD28101542
  • Cas Number : 227602-36-0
  • Molecular Weight : 284.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437471

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4834_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart