KEY428412430 Key Organics / BIONET

2-(2,6-dichlorophenyl)-2-methylpropanenitrile

  • MDL number MFCD11036576
  • Cas Number : 959139-99-2
  • Molecular Weight : 214.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 0.95
Storage temp. RT
Physical Form Colourless to Dark Yellow Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437472

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4835_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart