KEY428412431 Key Organics / BIONET

ethyl 2,4,6-trimethylpyrimidine-5-carboxylate

  • MDL number MFCD26405694
  • Cas Number : 90905-54-7
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point Oil
Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437473

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4836_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart