KEY428412432 Key Organics / BIONET

1-bromo-2-(2-bromoethyl)benzene

  • MDL number MFCD00027929
  • Cas Number : 1074-15-3
  • Molecular Weight : 263.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437474

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4837_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart