KEY428412433 Key Organics / BIONET

2-{spiro[2.2]pentan-1-yl}ethan-1-ol

  • MDL number MFCD28101543
  • Cas Number : 118757-50-9
  • Molecular Weight : 112.172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437475

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4838_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart