KEY428412438 Key Organics / BIONET

(2S)-5-methoxy-1,2,3,4-tetrahydronaphthalen-2-amine hydrochloride

  • MDL number MFCD08443937
  • Cas Number : 58349-17-0
  • Molecular Weight : 213.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/437480

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4843_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart