KEY428741905 Key Organics / BIONET

2-(2-methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane

  • MDL number MFCD28122661
  • Cas Number : 335343-05-0
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440197

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3059_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart