KEY428741906 Key Organics / BIONET

4,4,5,5-tetramethyl-2-[4-(4,4,5,5-tetramethyl-1,3-dioxolan-2-yl)phenyl]-1,3,2-dioxaborolane

  • MDL number MFCD22495442
  • Cas Number : 620595-02-0
  • Molecular Weight : 332.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440200

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3062_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart