KEY428741909 Key Organics / BIONET

5,5-dimethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborinane

  • MDL number MFCD28122664
  • Cas Number : 627906-96-1
  • Molecular Weight : 240.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440207

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3069_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart