KEY428741914 Key Organics / BIONET

2-(4-methoxyphenyl)-5,5-dimethyl-1,3,2-dioxaborinane

  • MDL number MFCD09909078
  • Cas Number : 213596-33-9
  • Molecular Weight : 220.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440202

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3064_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart