KEY428741915 Key Organics / BIONET

2-(3-ethenylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD18733869
  • Cas Number : 627525-99-9
  • Molecular Weight : 230.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store at 4
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440195

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3057_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart