KEY428741920 Key Organics / BIONET

1-{[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-1H-imidazole

  • MDL number MFCD14560559
  • Cas Number : 2246863-75-0
  • Molecular Weight : 284.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440208

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P302 + P352,P304 + P340,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3070_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart