KEY428741922 Key Organics / BIONET

2-(4-methoxynaphthalen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD22414634
  • Cas Number : 269410-16-4
  • Molecular Weight : 284.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-naphthyl ether
Inchi Code 1S/C17H21BO3/c1-16(2)17(3,4)21-18(20-16)14-10-11-15(19-5)13-9-7-6-8-12(13)14/h6-11H,1-5H3
Inchi Key PNGWDWZFEDHSPV-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440198

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3060_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart