KEY428741924 Key Organics / BIONET

3-bromofluoranthene

  • MDL number MFCD07787460
  • Cas Number : 13438-50-1
  • Molecular Weight : 281.152

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440212

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-3074_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart