KEY428741925 Key Organics / BIONET

5-bromo-2-methoxyphenyl acetate

  • MDL number MFCD27997554
  • Cas Number : 66037-04-5
  • Molecular Weight : 245.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440213

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-3075_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart