KEY428741928 Key Organics / BIONET

N-(3,4-difluorophenyl)-[1,2,4]triazolo[4,3-a]pyridine-8-sulfonamide

  • MDL number MFCD18804117
  • Cas Number : 1291486-52-6
  • Molecular Weight : 310.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Purple Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440216

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3300_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart