KEY428741931 Key Organics / BIONET

3-benzoyl-6,7-dimethoxy-1,4-dihydroquinolin-4-one

  • MDL number MFCD19104962
  • Cas Number : 1325303-43-2
  • Molecular Weight : 309.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440219

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3304_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart