KEY428741932 Key Organics / BIONET

2-amino-1-[4-(2-fluorophenyl)piperazin-1-yl]ethan-1-one hydrochloride

  • MDL number MFCD28041366
  • Cas Number : 1605772-63-1
  • Molecular Weight : 273.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440220

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3305_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart