KEY428741933 Key Organics / BIONET

2-[(2Z)-azepan-2-ylidene]-2-cyanoacetamide

  • MDL number MFCD20754284
  • Cas Number : 76140-88-0
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440221

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3306_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart