KEY428741937 Key Organics / BIONET

1-{3-[3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl]pyridin-2-yl}piperidine-4-carboxylic acid

  • MDL number MFCD18201580
  • Cas Number : 1260934-87-9
  • Molecular Weight : 384.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(3-(3-(3-chlorophenyl)-1,2,4-oxadiazol-5-yl)pyridin-2-yl)piperidine-4-carboxylic acid
Inchi Code 1S/C19H17ClN4O3/c20-14-4-1-3-13(11-14)16-22-18(27-23-16)15-5-2-8-21-17(15)24-9-6-12(7-10-24)19(25)26/h1-5,8,11-12H,6-7,9-10H2,(H,25,26)
Inchi Key PLAZEPLTYBSDJR-UHFFFAOYSA-N

Description

Purity 90%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440225

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3311_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart