KEY428741940 Key Organics / BIONET

3-[4-(benzyloxy)phenyl]-5-(chloromethyl)-1,2,4-oxadiazole

  • MDL number MFCD09034182
  • Cas Number : 937650-48-1
  • Molecular Weight : 300.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440228

Safety Information

signalword Warning
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404
hcodes H302,H312,H315,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3314_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart