KEY428741941 Key Organics / BIONET

(5-bromopyridin-2-yl)thiourea

  • MDL number MFCD15525454
  • Cas Number : 31430-38-3
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440229

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3315_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart