KEY428741942 Key Organics / BIONET

(Z)-2-[2-(4-fluorophenyl)-4-oxo-4H,5H-pyrazolo[1,5-a]pyrazin-5-yl]-N'-hydroxyethanimidamide

  • MDL number MFCD17780265
  • Cas Number : 1255791-17-3
  • Molecular Weight : 301.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440230

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3316_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart