KEY428741944 Key Organics / BIONET

[3-(methylsulfanyl)phenyl]urea

  • MDL number MFCD00175186
  • Cas Number : 85879-21-6
  • Molecular Weight : 182.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440232

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3319_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart