KEY428741952 Key Organics / BIONET

N-phenyl-3-(2-sulfanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide

  • MDL number MFCD14703430
  • Cas Number : 1193387-91-5
  • Molecular Weight : 295.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440240

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3333_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart