KEY428741955 Key Organics / BIONET

N-butyl-3-(2-sulfanylidene-2,3-dihydro-1H-imidazol-1-yl)benzamide

  • MDL number MFCD14703429
  • Cas Number : 1146290-03-0
  • Molecular Weight : 275.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-butyl-3-(2-sulfanyl-1H-imidazol-1-yl)benzamide
Inchi Code 1S/C14H17N3OS/c1-2-3-7-15-13(18)11-5-4-6-12(10-11)17-9-8-16-14(17)19/h4-6,8-10H,2-3,7H2,1H3,(H,15,18)(H,16,19)
Inchi Key TXRMBHKFVIBDRB-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440243

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3336_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart