KEY428741956 Key Organics / BIONET

(Z)-4-ethoxy-N'-hydroxy-3-methoxybenzene-1-carboximidamide

  • MDL number MFCD09939043
  • Cas Number : 885957-43-7
  • Molecular Weight : 210.233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440244

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3337_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart