KEY428741957 Key Organics / BIONET

2-(4-methoxybenzenesulfonyl)acetonitrile

  • MDL number MFCD00045627
  • Cas Number : 132276-87-0
  • Molecular Weight : 211.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440245

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3338_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart