KEY428741960 Key Organics / BIONET

ethyl 3-(morpholine-4-sulfonyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD18194701
  • Cas Number : 1108050-18-5
  • Molecular Weight : 289.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440248

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3342_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart