KEY428741967 Key Organics / BIONET

4-acetyl-N-(2,2-dimethoxyethyl)-3,5-dimethyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD14703586
  • Cas Number : 1232794-99-8
  • Molecular Weight : 268.313

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440255

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3349_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart