KEY428741970 Key Organics / BIONET

4,6-dimethyl-3-(3-phenyl-1,2,4-oxadiazol-5-yl)-1,2-dihydropyridin-2-one

  • MDL number MFCD14703532
  • Cas Number : 1223888-72-9
  • Molecular Weight : 267.288

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/440258

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3352_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart